6-Chloropiperonyl alcohol 98%

Leverantör: Thermo Fisher Scientific
Varning

L05972.06 L05972.14
L05972.06EA 940 SEK
L05972.06 L05972.14
6-Chloropiperonyl alcohol 98%
6-Chloropiperonyl alcohol

Purity: 98% CAS No.: 2591-25-5 Molecular Formula: C8H7ClO3 Formula Weight: 186.60 Melting Point: 68-72° Beilstein Reference: 150801 MDL No.: MFCD00051727

Formel: C₈H₇ClO₃
MW: 186.60 g/mol
Smältpunkt: 68…72 °C
Storage Temperature: Omgivande
MDL Number: MFCD00051727
CAS nummer: 2591-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR