Perfluoro(methyldecalin) mixture of 1- and 2-isomers (∼1:1) ≥85%, tech.
Leverantör: Thermo Scientific
Synonyms: Heptadecafluorodecahydro(trifluoromethyl)naphthalene, Perfluoro(methyldecahydronaphthalene)
L05862.14 L05862.22
L05862.14EA 1100 SEK
L05862.14 L05862.22
Perfluoro(methyldecalin) mixture of 1- and 2-isomers (∼1:1) ≥85%, tech.
Perfluoro(methyldecalin)
Purity: 85% CAS No.: 51294-16-7 Molecular Formula: C11F20 Formula Weight: 512.09 Refractive Index: 1.3170 Melting Point: -70° Boiling Point: ca 160° MDL No.: MFCD00014326
Formel: C₁₁F₂₀
MW: 512,09 g/mol
Kokpunkt: ∼160 °C
Smältpunkt: –70 °C
Densitet: 1,972
Storage Temperature: Omgivande
MDL Number: MFCD00014326
CAS nummer: 51294-16-7