N,N-Dimethylbutyramide 98%
Leverantör: Thermo Fisher Scientific
L05556.06
L05556.14
L05556.06EA
1775
SEK
L05556.06
L05556.14
N,N-Dimethylbutyramide 98%
N,N-Dimethylbutyramide
Purity: 98%
CAS No.: 760-79-2
Molecular Formula: C6H13NO
Formula Weight: 115.17
Refractive Index: 1.4425
Boiling Point: 80-82°/15mm
Beilstein Reference: 1742172
MDL No.: MFCD00015222
Formel:
C₆H₁₃NO Kokpunkt: 80…82 °C (15 mmHg) Storage Temperature: Omgivande |
MDL Number:
MFCD00015222 CAS nummer: 760-79-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...