Isophthalamide 98%

Leverantör: Thermo Scientific

L05137.06 L05137.14
L05137.06EA 650 SEK
L05137.06 L05137.14
Isophthalamide 98%
Isophthalamide

Purity: 98% CAS No.: 1740-57-4 Molecular Formula: C8H8N2O2 Formula Weight: 164.16 Melting Point: 281-282° Beilstein Reference: 2045544 MDL No.: MFCD00014801

Formel: C₈H₈N₂O₂
MW: 164,16 g/mol
Smältpunkt: 281…282 °C
Storage Temperature: Omgivande
MDL Number: MFCD00014801
CAS nummer: 1740-57-4
EINECS: 217-104-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR