(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: L-Cysteine ethyl ester hydrochloride, H-Cys-Oet.HCl, H-Cys-OEt·HCl
L04692.14 L04692.22
L04692.14EA 1200 SEK
L04692.14 L04692.22
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride ≥98%
(R)-Ethyl-2-amino-3-mercaptopropanoate hydrochloride
Purity: 98+% CAS No.: 868-59-7 Molecular Formula: C5H11NO2S.HCl Formula Weight: 185.67 Melting Point: 123-125° Beilstein Reference: 3562600 Optical Rotation: -12° (c=1 in 1N HCl) MDL No.: MFCD00012631
Formel: C₅H₁₁NO₂S·HCl
MW: 185,666 g/mol
Smältpunkt: 123…125 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012631
CAS nummer: 868-59-7
EINECS: 212-779-6