5-Oxohexanoic acid 97%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
Glurate, 5-Ketohexanoic Acid, 4-Acetylbutyric Acid
L03978.06
L03978.14
L03978.06EA
1125
SEK
L03978.06
L03978.14
5-Oxohexanoic acid 97%
5-Oxohexanoic acid
Purity: 97%
CAS No.: 3128-06-1
Molecular Formula: CH3CO(CH2)3CO2H
Formula Weight: 130.14
Flash Point:
Formel:
C₆H₁₀O₃ MW: 130,14 g/mol Kokpunkt: 274…275 °C Smältpunkt: 12…14 °C Densitet: 1,100 Flampunkt: >110 °C (230 °F) Storage Temperature: Omgivande |
MDL Number:
MFCD00004412 CAS nummer: 3128-06-1 EINECS: 221-511-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...