Pentamethylene sulphide 98%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Thiane, Tetrahydro-2H-thiopyran
L03063.03 L03063.06
L03063.03EA 730 SEK
L03063.03 L03063.06
Pentamethylene sulphide 98%
Pentamethylene sulphide
Purity: 98% CAS No.: 1613-51-0 Molecular Formula: C5H10S Formula Weight: 102.20 Flash Point: 21°(69°F) Refractive Index: 1.5060 Odour: Unpleasant Boiling Point: 140-142° Beilstein Reference: 102437 UN No.: UN1993 MDL No.: MFCD00006662
Formel: C₅H₁₀S
Kokpunkt: 140…142 °C
Densitet: 0,988
Flampunkt: 21 °C (69 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00006662
CAS nummer: 1613-51-0
EINECS: 216-561-1
ADR: 3,II