4-(Methylthio)toluene 97%
Leverantör: Thermo Fisher Scientific
Varning
L02933.06 L02933.14
L02933.06EA 1175 SEK
L02933.06 L02933.14
4-(Methylthio)toluene 97%
4-(Methylthio)toluene
Purity: 97% CAS No.: 623-13-2 Molecular Formula: C8H10S Formula Weight: 138.23 Flash Point: 85°(185°F) Refractive Index: 1.5730 Odour: Unpleasant Boiling Point: 81-82°/18mm Beilstein Reference: 1905733 UN No.: UN3334 MDL No.: MFCD00008560
Formel: C₈H₁₀S
Kokpunkt: 81…82 °C (18 mmHg)
Densitet: 1,029
Flampunkt: 85 °C (185 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00008560
CAS nummer: 623-13-2
EINECS: 210-773-8
ADR: 9,III