4-Hydroxy-3,5-diiodobenzoic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
L02595.09 L02595.18
L02595.09EA 1550 SEK
L02595.09 L02595.18
4-Hydroxy-3,5-diiodobenzoic acid 97%
4-Hydroxy-3,5-diiodobenzoic acid
Purity: 97% CAS No.: 618-76-8 Molecular Formula: C7H4I2O3 Formula Weight: 389.91 Melting Point: ca 275° dec. Beilstein Reference: 2212145 MDL No.: MFCD00016532
Formel: C₇H₄I₂O₃
Smältpunkt: ∼ 275 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00016532
CAS nummer: 618-76-8
EINECS: 210-562-0