2-Methylcyclohexylamine (cis- and trans- mixture) 97%
Leverantör: Thermo Fisher Scientific
Fara
L02285.09 L02285.18
L02285.09EA 625 SEK
L02285.09 L02285.18
2-Methylcyclohexylamine (cis- and trans- mixture) 97%
2-Methylcyclohexylamine (cis- and trans- mixture)
Purity: 97% CAS No.: 7003-32-9 Molecular Formula: C7H15N Formula Weight: 113.20 Flash Point: 21°(69°F) Refractive Index: 1.4565 Boiling Point: 149-150° Beilstein Reference: 2689073 UN No.: UN2733 MDL No.: MFCD00001492
Formel: C₇H₁₅N
Kokpunkt: 149…150 °C
Densitet: 0,858
Flampunkt: 21 °C (69 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00001492
CAS nummer: 7003-32-9
EINECS: 230-277-5
ADR: 3,II