2-(4-Isopropylphenoxy)acetic acid ≥98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
L01101.06
L01101.14
L01101.06EA
1450
SEK
L01101.06
L01101.14
2-(4-Isopropylphenoxy)acetic acid ≥98%
2-(4-Isopropylphenoxy)acetic acid
Purity: 98+%
CAS No.: 1643-16-9
Molecular Formula: C11H14O3
Formula Weight: 194.23
Melting Point: 85-87°
Beilstein Reference: 2261017
MDL No.: MFCD00014364
Formel:
C₁₁H₁₄O₃ Smältpunkt: 85…87 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00014364 CAS nummer: 1643-16-9 |