4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl 96% free radical
Leverantör: Thermo Scientific
Varning
Synonyms: TEMPONE, 4-Oxo-2,2,6,6-tetramethylpiperidinooxy, 4-Oxo-TEMPO
L01084.06 L01084.03
L01084.06EA 4600 SEK
L01084.06 L01084.03
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl 96% free radical
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl
Purity: 96% CAS No.: 2896-70-0 Molecular Formula: C9H16NO2 Formula Weight: 170.23 Melting Point: 36-40° Beilstein Reference: 1818579 MDL No.: MFCD00006187
Formel: C₉H₁₆NO₂
MW: 170,23 g/mol
Smältpunkt: 36…40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00006187
CAS nummer: 2896-70-0
EINECS: 220-778-7