2-(8-Bromooctyloxy)tetrahydropyran 90%, tech.
Leverantör: Thermo Fisher Scientific
L01081.06 L01081.14
L01081.06EA 1250 SEK
L01081.06 L01081.14
2-(8-Bromooctyloxy)tetrahydropyran 90%, tech.
2-(8-Bromooctyloxy)tetrahydropyran
Purity: 90% CAS No.: 50816-20-1 Molecular Formula: C13H25BrO2 Formula Weight: 293.25 Flash Point: 110°(230°F) Refractive Index: 1.4770 Boiling Point: 130-132°/0.8mm Beilstein Reference: 1306644 MDL No.: MFCD00014648
Formel: C₁₃H₂₅BrO₂
MW: 293.25 g/mol
Kokpunkt: 130…132 °C (0.8 mmHg)
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00014648
CAS nummer: 50816-20-1
EINECS: 256-786-2