Natriumdodecylsulfat 99% för molekylärbiologi
Leverantör: Thermo Fisher Scientific
|
Fara
|
Purity: 99%
CAS No.: 151-21-3
Molecular Formula: C12H25NaO4S
Formula Weight: 288.38
Physical Form: Solid
Appearance: White
Melting Point: ca 206°
Solubility: Soluble in water
Beilstein Reference: 3599286
UN No.: UN2926
MDL No.: MFCD00036175
Formel:
CH₃(CH₂)₁₁OSO₃Na Smältpunkt: ∼206 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00036175 CAS nummer: 151-21-3 EINECS: 205-788-1 UN: 1325 ADR: 4.1,III Merck Index: 14,8636 |