Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 80 nm (Abs. 457 nm)
Leverantör: Thermo Fisher Scientific
J67252.AC J67252.AE
J67252.ACEA 2225 SEK
J67252.AC J67252.AE
Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 80 nm (Abs. 457 nm)
Silver
CAS No.: 7440-22-4 Storage Temperature: 2°C to 8°C Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Yellow Odour: Odourless MDL No.: MFCD00003397
Formel: Ag
MW: 107.87 g/mol
Kokpunkt: 2212
Smältpunkt: 962
Storage Temperature: Kylskåp
MDL Number: MFCD00003397
CAS nummer: 7440-22-4
Merck Index: 12,08647