orto-Borsyra ≥99.5% för elektrofores
Leverantör: Thermo Fisher Scientific
|
Fara
|
Purity: 99.5% min
CAS No.: 10043-35-3
Molecular Formula: BH3O3
Formula Weight: 61.83
Physical Form: Powder
Density: 1.435
Appearance: White
Odour: Odourless
pH: 3.8-4.8
Melting Point: ca 185° dec
Beilstein Reference: 1697939
MDL No.: MFCD00011337
Formel:
B(OH)₃ Smältpunkt: ∼ 185 °C (dec.) Densitet: 1,435 Storage Temperature: Omgivande |
MDL Number:
MFCD00011337 CAS nummer: 10043-35-3 EINECS: 233-139-2 Merck Index: 14,1336 |