N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
Leverantör: Thermo Fisher Scientific
Varning
J66265.06 J66265.14
J66265.06EA 2775 SEK
J66265.06 J66265.14
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulphonate(sulphobetaine-12, SB-12)
Applications: For protein solubilization
Applications: For protein solubilization
Formel: C₁₇H₃₇NO₃S
Flampunkt: 110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00036909
CAS nummer: 14933-08-5
EINECS: 239-002-3
UN: 3261
ADR: 8,II