Cefixime trihydrate
Leverantör: Thermo Fisher Scientific
Fara
J66094.MC J66094.03
J66094.MCEA 1975 SEK
J66094.MC J66094.03
Cefixime trihydrate
Cefixime trihydrate
A broad spectrum third generation cephalosporin.
  • Air and Light Sensitive
Pale yellow powder
Applications: A broad spectrum third generation cephalosporin
Soluble in water at 55.1 µg/mL
Formel: C₁₆H₂₁N₅O₁₀S₂
MW: 507.489 g/mol
Smältpunkt: 218…225 °C
Storage Temperature: Kylskåp
MDL Number: MFCD03788802
CAS nummer: 125110-14-7