Fluconazole 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-(2,4-Difluorophenyl)-1,3-bis(1H-1,2,4-triazol-1-yl)propan-2-ol, 2-(2,4-Difluorophenyl)-1,3-di(1H-1,2,4-triazol-1-yl)propan-2-ol
J62015.03 J62015.06 J62015.14
J62015.03EA 2500 SEK
J62015.03 J62015.06 J62015.14
Fluconazole 99%
Fluconazole
Purity: 99% CAS No.: 86386-73-4 Molecular Formula: C13H12F2N6O Formula Weight: 306.27 Physical Form: Crystalline powder Density: 1.05 Appearance: White to off-white Melting Point: 138-140° MDL No.: MFCD00274549
Formel: C₁₃H₁₂F₂N₆O
MW: 306,28 g/mol
Smältpunkt: 138…140 °C
Densitet: 1,05
Storage Temperature: Omgivande
MDL Number: MFCD00274549
CAS nummer: 86386-73-4
Merck Index: 14,4122