Nicorandil ≥98%

Leverantör: Thermo Fisher Scientific
Fara

Synonyms: N-[2-(Nitrooxy)ethyl]pyridine-3-carboxamide, 2-(Pyridine-3-carboxamido)ethyl Nitrate, 2-(Pyridine-3-carbonylamino)ethyl nitrate

J60879.MD J60879.03
J60879.MDEA 1700 SEK
J60879.MD J60879.03
Nicorandil ≥98%
Nicorandil

Purity: 98+% CAS No.: 65141-46-0 Molecular Formula: C8H9N3O4 Formula Weight: 211.17 Physical Form: Powder Appearance: White to off-white Melting Point: 92-93° MDL No.: MFCD00186520


Applications: A vasodilator that activates ATP-sensitive potassium channels

Formel: C₈H₉N₃O₄
Smältpunkt: 92…93 °C
Storage Temperature: Omgivande
MDL Number: MFCD00186520
CAS nummer: 65141-46-0
EINECS: 265-514-1
Merck Index: 14,6521

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR