Orange G 6X loading dye, glycerol based
Leverantör: Thermo Fisher Scientific
Synonyms: CI 16230
J60562.ACEA 1525 SEK
J60562.AC J60562.AD
Orange G 6X loading dye, glycerol based
Orange G
DNA loading dye used to prepare markers and samples for loading on agarose or polyacrylamide gels in molecular biology protocols.
  • Recommended for small molecules
The 6X Orange DNA Loading Dye is used to prepare DNA markers and samples for loading on agarose or polyacrylamide gels. It is recommended for the analysis of small DNA molecules.
Contains: 0.25% Orange-G and 30% glycerol in autoclaved DI water
Formel: C₁₆H₁₀N₂Na₂O₇S₂
MW: 452.38 g/mol
Smältpunkt: 141
Storage Temperature: Kylskåp
MDL Number: MFCD00012457
CAS nummer: 1936-15-8
Order Now
SPECIFIKATIONER
Environmentally Preferable