TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98% för molekylärbiologi
Leverantör: Thermo Scientific
Fara
Synonyms: TCEP HCl
J60316.06 J60316.09 J60316.3S
J60316.06EA 4025 SEK
J60316.06 J60316.09 J60316.3S
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride) 98% för molekylärbiologi
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)
Applications: For reducing disulfide bonds in peptides and proteins.
Applications: For reducing disulfide bonds in peptides and proteins.
Formel: C₉H₁₆ClO₆P
MW: 286,65 g/mol
Smältpunkt: 177…178 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00145469
CAS nummer: 51805-45-9
UN: 3261
ADR: 8,II
Merck Index: 14,9085