4,6-Dichloro-o-cresol 98%
Leverantör: Thermo Scientific
Fara
Synonyms: 3,5-Dichloro-2-hydroxytoluene, 2,4-Dichloro-6-methylphenol, 1,3-Dichloro-4-hydroxy-5-methylbenzene
H64312.03 H64312.06
H64312.03EA 1250 SEK
H64312.03 H64312.06
4,6-Dichloro-o-cresol 98%
4,6-Dichloro-o-cresol
Purity: 98% CAS No.: 1570-65-6 Molecular Formula: C7H6Cl2O Formula Weight: 177.02 Physical Form: Solid Appearance: Brown Melting Point: 52-56° Boiling Point: 225° lit. Beilstein Reference: 1864865 UN No.: UN3077 MDL No.: MFCD00019988
Formel: C₇H₆Cl₂O
MW: 177,03 g/mol
Kokpunkt: 225 °C (lit.)
Smältpunkt: 52…56 °C
Storage Temperature: Omgivande
MDL Number: MFCD00019988
CAS nummer: 1570-65-6
EINECS: 216-382-9
ADR: ,