L-1-Naphthylalanine 95%
Leverantör: Thermo Scientific
Synonyms:
H-1-Nal-OH, 3-(1-Naphthyl)-L-alanine
H63586.03
H63586.06
H63586.03EA
1525
SEK
H63586.03
H63586.06
L-1-Naphthylalanine 95%
L-1-Naphthylalanine
Purity: 95%
CAS No.: 55516-54-6
Molecular Formula: C13H13NO2
Formula Weight: 215.25
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 4352833
MDL No.: MFCD00066086
Formel:
C₁₃H₁₃NO₂ MW: 215,252 g/mol |
MDL Number:
MFCD00066086 CAS nummer: 55516-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...