Silverklorid 4,9% i vatten, nanoparticle dispersion <200nm APS NW
Leverantör: Thermo Scientific
47280.22 47280.36
47280.22EA 5400 SEK
47280.22 47280.36
Silverklorid 4,9% i vatten, nanoparticle dispersion <200nm APS NW
Silver chloride
CAS No.: 7783-90-6 Molecular Formula: AgCl Formula Weight: 143.32 Storage Temperature: 2°C to 8°C Physical Form: Liquid Density: 1.038 Appearance: White opaque Odour: Odourless Merck Reference: 14,8509 UN No.: 3082 MDL No.: MFCD00003399
Formel: AgCl
MW: 143,32 g/mol
Densitet: 1,038
Storage Temperature: Kylskåp
MDL Number: MFCD00003399
CAS nummer: 7783-90-6
EINECS: 232-033-3
UN: 3082
ADR: 9,III
Merck Index: 14,8509