Aluminium oxide activated basic HPLC_Flash
Leverantör: Thermo Fisher Scientific
45949.A1 45949.30
45949.A1EA 5575 SEK
45949.A1 45949.30
Aluminium oxide activated basic HPLC_Flash
Aluminium oxide activated basic
CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 18-32 Micron APS Powder Refractive Index: 1.765 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424
Formel: Al₂O₃
MW: 101.96 g/mol
Kokpunkt: 2980 °C
Smältpunkt: 2045 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003424
CAS nummer: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356