3-(2-Pyridyl)propionic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 3-(2-Pyridyl)propionic Acid
H58619.03 H58619.06
H58619.03EA 3275 SEK
H58619.03 H58619.06
3-(2-Pyridyl)propionic acid 97%
3-(2-Pyridyl)propionic acid
Purity: 97% CAS No.: 15197-75-8 Molecular Formula: C8H9NO2 Formula Weight: 151.16 MDL No.: MFCD00995159
Formel: C₈H₉NO₂
MW: 151.16 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00995159
CAS nummer: 15197-75-8