(R)-Pyrrolidin-3-ylmethanol 95%
Leverantör: Thermo Scientific
Varning
H57871.MD H57871.03
H57871.MDEA 3100 SEK
H57871.MD H57871.03
(R)-Pyrrolidin-3-ylmethanol 95%
(R)-Pyrrolidin-3-ylmethanol
Purity: 95% CAS No.: 110013-18-8 Molecular Formula: C5H11NO Formula Weight: 101.15 Storage Temperature: 2°C to 8°C Boiling Point: 173-177° MDL No.: MFCD09607969
Formel: C₅H₁₁NO
MW: 101,15 g/mol
Kokpunkt: 173…177 °C
Densitet: 0,979
MDL Number: MFCD09607969
CAS nummer: 110013-18-8