4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one 97%
Leverantör: Thermo Scientific
Fara
H55735.MD H55735.03 H55735.06
H55735.MDEA 610 SEK
H55735.MD H55735.03 H55735.06
4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one 97%
4-(Chloromethyl)-5-methyl-1,3-dioxol-2-one
Purity: 97% CAS No.: 80841-78-7 Molecular Formula: C5H5ClO3 Formula Weight: 148.55 Flash Point: 129°(264°F) Boiling Point: 90°/2mm Beilstein Reference: 4798926 MDL No.: MFCD07787494
Formel: C₅H₅ClO₃
MW: 148,55 g/mol
Kokpunkt: >90 °C (2 mmHg)
Flampunkt: 129 °C (264 °F)
Storage Temperature: Omgivande
MDL Number: MFCD07787494
CAS nummer: 80841-78-7