1,2,3,4-Tetrahydroquinoxaline 98%
Leverantör: Thermo Fisher Scientific
|
Varning
|
H55473.MD
H55473.03
H55473.06
H55473.MDEA
1475
SEK
H55473.MD
H55473.03
H55473.06
1,2,3,4-Tetrahydroquinoxaline 98%
1,2,3,4-Tetrahydroquinoxaline
Purity: 98%
CAS No.: 3476-89-9
Molecular Formula: C8H10N2
Formula Weight: 134.18
Melting Point: 92-98°
MDL No.: MFCD00047564
Formel:
C₈H₁₀N₂ MW: 134.18 g/mol Smältpunkt: 92…98 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00047564 CAS nummer: 3476-89-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...