2-(Methylamino)acetonitrile hydrochloride 98%
Leverantör: Thermo Scientific
Fara
H55215.06 H55215.14
H55215.06EA 890 SEK
H55215.06 H55215.14
2-(Methylamino)acetonitrile hydrochloride 98%
2-(Methylamino)acetonitrile hydrochloride
Purity: 99% CAS No.: 25808-30-4 Molecular Formula: C3H6N2.HCl Formula Weight: 106.55 Melting Point: 103-106° Beilstein Reference: 3908975 MDL No.: MFCD00012587
Formel: C₃H₇ClN₂
MW: 106,55 g/mol
Smältpunkt: 103…106 °C
Storage Temperature: Omgivande
MDL Number: MFCD00012587
CAS nummer: 25808-30-4
EINECS: 247-277-6