3-Ethyl-4-methyl-3-pyrrolin-2-one 98%
Leverantör: Thermo Scientific
Synonyms: 3-Ethyl-4-methyl-2,5-dihydro-1H-pyrrol-2-one, 1,5-Dihydro-3-ethyl-4-methyl-2H-pyrrol-2-one
H55065.03 H55065.06
H55065.03EA 1125 SEK
H55065.03 H55065.06
3-Ethyl-4-methyl-3-pyrrolin-2-one 98%
3-Ethyl-4-methyl-3-pyrrolin-2-one
Purity: 98% CAS No.: 766-36-9 Molecular Formula: C7H11NO Formula Weight: 125.17 Melting Point: 100-104° MDL No.: MFCD00173861
Formel: C₇H₁₁NO
MW: 125,17 g/mol
Smältpunkt: 100…104 °C
Storage Temperature: Omgivande
MDL Number: MFCD00173861
CAS nummer: 766-36-9