2,2-Bis[4-(4-aminophenoxy)phenyl]propane 98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4,4'-Isopropylidenebis[(4-aminophenoxy)benzene], 4,4'-((Propane-2,2-diylbis(4,1-phenylene))bis(oxy))dianiline
H55057.14 H55057.22
H55057.14EA 1125 SEK
H55057.14 H55057.22
2,2-Bis[4-(4-aminophenoxy)phenyl]propane 98%
2,2-Bis[4-(4-aminophenoxy)phenyl]propane
Purity: 98% CAS No.: 13080-86-9 Molecular Formula: C27H26N2O2 Formula Weight: 410.52 Melting Point: 127-130° MDL No.: MFCD00039152
Formel: C₂₇H₂₆N₂O₂
MW: 410,52 g/mol
Smältpunkt: 127…130 °C
Storage Temperature: Omgivande
MDL Number: MFCD00039152
CAS nummer: 13080-86-9
EINECS: 235-985-8