tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Bis-Boc-pyrazolocarboxamidine, 1-[N,N'-Bis(tert-butoxycarbonyl)carbamimidoyl]-1H-pyrazole, N,N'-Di-BOC-1H-pyrazole-1-carboxamidine, N,N'-Di-Boc-1H-pyrazole-1-carboximidamide, 1H-Pyrazole-1-carboxamidine, N,N'-Bis-BOC protected, N,N'-Bis(tert-butoxycarbonyl)-1H-pyrazole-1-carboximidamide
H54565.03 H54565.06 H54565.14
H54565.03EA 1250 SEK
H54565.03 H54565.06 H54565.14
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate ≥98%
tert-Butyl (((tert-butoxycarbonyl)amino)(1H-pyrazol-1-yl)methylene)carbamate
Purity: 98+% CAS No.: 152120-54-2 Molecular Formula: C14H22N4O4 Formula Weight: 310.35 Melting Point: 86-90° MDL No.: MFCD01075122
Formel: C₁₄H₂₂N₄O₄
MW: 310,354 g/mol
Smältpunkt: 86…90 °C
Storage Temperature: Omgivande
MDL Number: MFCD01075122
CAS nummer: 152120-54-2