6-Bromo-1-indanone 97%
Leverantör: Thermo Scientific
H54003.03 H54003.06
H54003.03EA 1625 SEK
H54003.03 H54003.06
6-Bromo-1-indanone 97%
6-Bromo-1-indanone
Purity: 97% CAS No.: 14548-39-1 Molecular Formula: C9H7BrO Formula Weight: 211.06 Melting Point: 110° MDL No.: MFCD02179286
Formel: C₉H₇BrO
MW: 211,06 g/mol
Smältpunkt: 110 °C
Storage Temperature: Omgivande
MDL Number: MFCD02179286
CAS nummer: 14548-39-1