5,7-Difluoro-1-indanone 97%
Leverantör: Thermo Scientific
Fara
H54002.03 H54002.06
H54002.03EA 1650 SEK
H54002.03 H54002.06
5,7-Difluoro-1-indanone 97%
5,7-Difluoro-1-indanone
Purity: 97% CAS No.: 84315-25-3 Molecular Formula: C9H6F2O Formula Weight: 168.14 Melting Point: 81° MDL No.: MFCD09908161
Formel: C₉H₆F₂O
MW: 168,14 g/mol
Smältpunkt: 81 °C
Storage Temperature: Omgivande
MDL Number: MFCD09908161
CAS nummer: 84315-25-3