1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: α-[2-(Trimethylsilyl)ethynyl]benzenemethanol
H53426.06 H53426.14
H53426.06EA 1925 SEK
H53426.06 H53426.14
1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol 97%
1-Phenyl-3-(trimethylsilyl)-2-propyn-1-ol
Purity: 97% CAS No.: 89530-34-7 Molecular Formula: C12H16OSi Formula Weight: 204.34 Flash Point: 110°(230°F) Refractive Index: 1.5193 MDL No.: MFCD05864336
Formel: C₁₂H₁₆OSi
Densitet: 0,961
Flampunkt: 110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD05864336
CAS nummer: 89530-34-7