DL-Isoserine 98%
Leverantör: Thermo Scientific
Synonyms: H-DL-IsoSer-OH, DL-3-Amino-2-hydroxypropionic Acid
H53412.03 H53412.06
H53412.03EA 5125 SEK
H53412.03 H53412.06
DL-Isoserine 98%
DL-Isoserine
Purity: 98% CAS No.: 565-71-9 Molecular Formula: C3H7NO3 Formula Weight: 105.09 Melting Point: 235° dec. Beilstein Reference: 1721413 MDL No.: MFCD00008138
Formel: C₃H₇NO₃
MW: 105,09 g/mol
Smältpunkt: 235 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00008138
CAS nummer: 565-71-9