1-(1-Adamantyl)ethylamine hydrochloride 98%
Leverantör: Thermo Scientific
Fara
Synonyms: Rimantadine Hydrochloride, alpha-Methyl-1-adamantanemethylamine Hydrochloride
H53377.06 H53377.14
H53377.06EA 2125 SEK
H53377.06 H53377.14
1-(1-Adamantyl)ethylamine hydrochloride 98%
1-(1-Adamantyl)ethylamine hydrochloride
Purity: 99% CAS No.: 1501-84-4 Molecular Formula: C12H21N.HCl Formula Weight: 215.77 Melting Point: 375° MDL No.: MFCD00072023
Formel: C₁₂H₂₁N
MW: 215,77 g/mol
Smältpunkt: 375 °C
Storage Temperature: Omgivande
MDL Number: MFCD00072023
CAS nummer: 1501-84-4
Merck Index: 14,8224