Fullerene nanotube multi-walled ≤8 nm OD, 2-5 nm ID, Length: 0.5-2 micron
Leverantör: Thermo Scientific
Varning
Synonyms: Fullerene carbon nanotube
44790.01 44790.03 44790.06
44790.01EA 2125 SEK
44790.01 44790.03 44790.06
Fullerene nanotube multi-walled ≤8 nm OD, 2-5 nm ID, Length: 0.5-2 micron
Fullerene nanotube
Smältpunkt: > 280 °C
Densitet: ∼1,65 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00133992
CAS nummer: 308068-56-6
UN: 1325
ADR: 4.1,III
Merck Index: 13,01447