Boc-trans-4-amino-L-proline methyl ester hydrochloride salt 97%
Leverantör: Thermo Fisher Scientific
Synonyms: (2S,4R)-1-tert-Butyl 2-methyl 4-aminopyrrolidine-1,2-dicarboxylate hydrochloride, Methyl (2S,4R)-4-aminopyrrolidine-2-carboxylate hydrochloride, N1-BOC protected
H52566.03 H52566.06
H52566.03EA 3525 SEK
H52566.03 H52566.06
Boc-trans-4-amino-L-proline methyl ester hydrochloride salt 97%
Boc-trans-4-amino-L-proline methyl ester hydrochloride salt
Purity: 97% CAS No.: 334999-32-5 Molecular Formula: C11H20N2O4.HCl Formula Weight: 280.75 MDL No.: MFCD03787929
Formel: C₁₁H₂₁ClN₂O₄
MW: 280.75 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD03787929
CAS nummer: 334999-32-5