(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine 97%
Leverantör: Thermo Scientific
Fara
Synonyms: tert-Butyl (R)-2-(Aminomethyl)-1-pyrrolidinecarboxylate, (R)-2-(Aminomethyl)-1-Boc-pyrrolidine, (R)-2-(Aminomethyl)-1-(tert-butoxycarbonyl)pyrrolidine, (R)-2-(Aminomethyl)-1-pyrrolidinecarboxylic acid tert-Butyl ester
H52419.03 H52419.06
H52419.03EA 2675 SEK
H52419.03 H52419.06
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine 97%
(R)-2-(Aminomethyl)-1-N-boc-pyrrolidine
Purity: 97% CAS No.: 259537-92-3 Molecular Formula: C10H20N2O2 Formula Weight: 200.28 Boiling Point: 98-110°/1mm Optical Rotation: +47° (c=1 in chloroform) UN No.: UN2735 MDL No.: MFCD03419256
Formel: C₁₀H₂₀N₂O₂
MW: 200,28 g/mol
Kokpunkt: 98…110 °C (1 mmHg)
Storage Temperature: Omgivande
MDL Number: MFCD03419256
CAS nummer: 259537-92-3
ADR: 8,III