2-Bromo-3-(methoxymethoxy)pyridine 96%
Leverantör: Thermo Fisher Scientific
Varning
H52324.03 H52324.06
H52324.03EA 5550 SEK
H52324.03 H52324.06
2-Bromo-3-(methoxymethoxy)pyridine 96%
2-Bromo-3-(methoxymethoxy)pyridine
Purity: 96% CAS No.: 162271-10-5 Molecular Formula: C7H8BrNO2 Formula Weight: 218.05 Melting Point: 35-37° MDL No.: MFCD17015031
Formel: C₇H₈BrNO₂
MW: 218.05 g/mol
Smältpunkt: 35…37 °C
Storage Temperature: Omgivande
MDL Number: MFCD17015031
CAS nummer: 162271-10-5