5-Bromo-1,3-dimethyl-2-pyridone 97%
Leverantör: Thermo Scientific
Fara
Synonyms: 5-Bromo-1,3-dimethylpyridin-2(1H)-one
H52279.03 H52279.06
H52279.03EA 3625 SEK
H52279.03 H52279.06
5-Bromo-1,3-dimethyl-2-pyridone 97%
5-Bromo-1,3-dimethyl-2-pyridone
Purity: 97% CAS No.: 51417-13-1 Molecular Formula: C7H8BrNO Formula Weight: 202.05 Melting Point: 105-106°C MDL No.: MFCD16658881
Formel: C₇H₈BrNO
MW: 202,05 g/mol
Smältpunkt: 105…106 °C
Storage Temperature: Omgivande
MDL Number: MFCD16658881
CAS nummer: 51417-13-1