Fmoc-L-β-homoglutamic acid-d-t-butyl ester 95%
Leverantör: Thermo Scientific
Synonyms: Fmoc-β-HoGlu(OtBu)-OH, Fmoc-β-HomoGlu(OtBu)-OH
H52188.MD H52188.03
H52188.MDEA 3600 SEK
H52188.MD H52188.03
Fmoc-L-β-homoglutamic acid-d-t-butyl ester 95%
Fmoc-L-beta-homoglutamic acid-d-t-butyl ester
Purity: 95% CAS No.: 203854-49-3 Molecular Formula: C25H29NO6 Formula Weight: 439.51 Melting Point: 115-120° Beilstein Reference: 8019724 MDL No.: MFCD01863053
Formel: C₂₅H₂₉NO₆
MW: 439,51 g/mol
Smältpunkt: 115…120 °C
Storage Temperature: Omgivande
MDL Number: MFCD01863053
CAS nummer: 203854-49-3