2-Benzyl-L-proline hydrochloride 95%

Leverantör: Thermo Fisher Scientific

H52080.MD H52080.03
H52080.MDEA 3700 SEK
H52080.MD H52080.03
2-Benzyl-L-proline hydrochloride 95%
2-Benzyl-L-proline hydrochloride

Purity: 95% CAS No.: 86116-84-9 Molecular Formula: C12H15NO2.HCl Formula Weight: 241.72 Melting Point: 270-272° Optical Rotation: -33° (c=1 in 1N HCl) MDL No.: MFCD16660381

Formel: C₁₂H₁₅NO₂·HCl
Smältpunkt: 270…272 °C
Storage Temperature: Omgivande
MDL Number: MFCD16660381
CAS nummer: 86116-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR