Fmoc-L-Phe(4-Br)-OH 95%
Leverantör: Thermo Fisher Scientific
H51978.03
H51978.06
H51978.03EA
2250
SEK
H51978.03
H51978.06
Fmoc-L-Phe(4-Br)-OH 95%
Fmoc-L-Phe(4-Br)-OH
Purity: 95%
CAS No.: 198561-04-5
Molecular Formula: C24H20BrNO4
Formula Weight: 466.33
Melting Point: 165-168°
MDL No.: MFCD00273460
Formel:
C₂₄H₂₀BrNO₄ Smältpunkt: 165…168 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00273460 CAS nummer: 198561-04-5 |