2-Chloro-5,5-dimethyl-1,3-cyclohexanedione 98%
Leverantör: Thermo Scientific
Varning
H51035.MD H51035.03
H51035.MDEA 2375 SEK
H51035.MD H51035.03
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione 98%
2-Chloro-5,5-dimethyl-1,3-cyclohexanedione
Purity: 98% CAS No.: 7298-89-7 Molecular Formula: C8H11ClO2 Formula Weight: 174.62 Melting Point: 161-164° Beilstein Reference: 2326749 MDL No.: MFCD00010502
Formel: C₈H₁₁ClO₂
MW: 174,62 g/mol
Smältpunkt: 161…164 °C
Storage Temperature: Omgivande
MDL Number: MFCD00010502
CAS nummer: 7298-89-7