4-(Pyrrolidin-3-yl)morpholine 97%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: 4-(3-Pyrrolidinyl)morpholine
H51002.03 H51002.MD
H51002.03EA 5150 SEK
H51002.03 H51002.MD
4-(Pyrrolidin-3-yl)morpholine 97%
4-(Pyrrolidin-3-yl)morpholine
Purity: 97% CAS No.: 53617-37-1 Molecular Formula: C8H16N2O Formula Weight: 156.23 UN No.: UN2735 MDL No.: MFCD06740352
Formel: C₈H₁₆N₂O
MW: 156.229 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD06740352
CAS nummer: 53617-37-1
ADR: 8 III