1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde 98%
Leverantör: Thermo Fisher Scientific
Varning
H50225.03 H50225.06
H50225.03EA 2025 SEK
H50225.03 H50225.06
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde 98%
1,3,5-Trimethyl-1H-pyrazole-4-carbaldehyde
Purity: 99% CAS No.: 2644-93-1 Molecular Formula: C7H10N2O Formula Weight: 138.17 Melting Point: 80-83° Boiling Point: 132°/12mm MDL No.: MFCD00159623
Formel: C₇H₁₀N₂O
MW: 138.17 g/mol
Kokpunkt: 132 °C (12 mmHg)
Smältpunkt: 80…83 °C
Storage Temperature: Omgivande
MDL Number: MFCD00159623
CAS nummer: 2644-93-1